Invitrogen
Manufacturer Code:PA599118
Catalog # PIPA599118
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit mouse Grx1 (full length) protein purified from E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glutaredoxin (thioltransferase); glutaredoxin 1 (thioltransferase); Glutaredoxin-1; Thioltransferase-1; TTase-1
Gene Aliases: C86710; D13Wsu156e; Glrx; Glrx1; Grx; Grx1; TTase
UniProt ID: (Mouse) Q9QUH0, (Rat) Q9ESH6
Entrez Gene ID: (Mouse) 93692, (Rat) 64045
Molecular Function: oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.