Invitrogen
Manufacturer Code:PA592389
Catalog # PIPA592389
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-106 of human GLRX |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glutaredoxin (thioltransferase); glutaredoxin 1 (thioltransferase); Glutaredoxin-1; thioltransferase; Thioltransferase-1; TTase-1
Gene Aliases: C86710; D13Wsu156e; GLRX; Glrx1; GRX; GRX1; TTase
UniProt ID: (Human) P35754, (Mouse) Q9QUH0, (Rat) Q9ESH6
Entrez Gene ID: (Human) 2745, (Mouse) 93692, (Rat) 64045
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.