Invitrogen
Manufacturer Code:PA570034
Catalog # PIPA570034
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human GLRX |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glrx Glrx1 Glutaredoxin Glutaredoxin1 Grx 1 Grx1 MGC117; glutaredoxin (thioltransferase); glutaredoxin 1 (thioltransferase); Glutaredoxin-1; thiol disulfide oxidoreductase; Thioltransferase; Thioltransferase-1; thioltransferase; TTase; TTase; TTase-1
Gene Aliases: C86710; D13Wsu156e; GLRX; GLRX1; GLRXL; GRX; GRX1; TTase; TTase-1; TTF
UniProt ID: (Human) P35754, (Pig) P12309, (Bovine) P10575, (Mouse) Q9QUH0, (Rat) Q9ESH6
Entrez Gene ID: (Human) 2745, (Pig) 397463, (Bovine) 515416, (Sheep) 100147797, (Horse) 100064937, (Rabbit) 100344913, (Mouse) 93692, (Rat) 64045, (Guinea pig) 100713710
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.