Invitrogen
Manufacturer Code:PA547451
Catalog # PIPA547451
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit mouse Glutaredoxin 1 Met1-Leu107 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Glrx Glrx1 Glutaredoxin Glutaredoxin1 Grx 1 Grx1 MGC117; glutaredoxin (thioltransferase); glutaredoxin 1 (thioltransferase); Glutaredoxin-1; Thioltransferase-1; TTase-1
Gene Aliases: C86710; D13Wsu156e; Glrx; Glrx1; Grx; Grx1; TTase
UniProt ID: (Mouse) Q9QUH0
Entrez Gene ID: (Mouse) 93692
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.