Proteintech
Manufacturer Code:158041AP150UL
Catalog # 50-173-0492
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GLRX Fusion Protein Ag8535 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GLRX Glutaredoxin 1 GRX GRX1 Thioltransferase 1 TTase 1; glutaredoxin (thioltransferase); glutaredoxin 1 (thioltransferase); Glutaredoxin-1; thioltransferase; Thioltransferase-1; TTase-1
Gene Aliases: C86710; D13Wsu156e; GLRX; Glrx1; GRX; GRX1; TTase
UniProt ID: (Human) P35754, (Mouse) Q9QUH0, (Rat) Q9ESH6
Entrez Gene ID: (Human) 2745, (Mouse) 93692, (Rat) 64045
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.