Invitrogen
Manufacturer Code:PA539191
Catalog # PIPA539191
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the internal region of human GLRB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glycine receptor 58 kDa subunit; Glycine receptor subunit beta; Glycine receptor subunit beta Glycine receptor 58 kDa subunit GlyR beta GlyR beta; glycine receptor, beta subunit; inhibitory glycine receptor
Gene Aliases: AI853901; GLRB; Glyrb; HKPX2; spa; spastic
UniProt ID: (Human) P48167, (Mouse) P48168, (Rat) P20781
Entrez Gene ID: (Human) 2743, (Mouse) 14658, (Rat) 25456
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.