Invitrogen
Manufacturer Code:PA5101610
Catalog # PIPA5101610
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human GLRB corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI853901 Glrb glycine receptor glycine receptor 58 kDa subunit glycine receptor beta glycine receptor subunit beta glycine receptor beta glycine receptor beta subunit GlyR beta Glyrb HKPX2 inhibitory glycine receptor spa spastic; Glycine receptor 58 kDa subunit; Glycine receptor subunit beta; glycine receptor, beta subunit; inhibitory glycine receptor
Gene Aliases: AI853901; GLRB; Glyrb; HKPX2; spa; spastic
UniProt ID: (Human) P48167, (Mouse) P48168, (Rat) P20781
Entrez Gene ID: (Human) 2743, (Mouse) 14658, (Rat) 25456
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.