Invitrogen
Manufacturer Code:PA577444
Catalog # PIPA577444
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)KEDVKSGLKGSQPMS corresponding to amino acid residues 28-42 of mouse Glycine Receptor alpha4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glycine Receptor alpha4; Glycine receptor subunit alpha-4; glycine receptor, alpha 4 subunit
Gene Aliases: Glra4; RGD1564103
UniProt ID: (Mouse) Q61603
Entrez Gene ID: (Human) 441509, (Mouse) 14657, (Rat) 367912
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.