Invitrogen
Manufacturer Code:PA590611
Catalog # PIPA590611
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 350-420 of human GLRA2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glycine receptor alpha 2 subunit; Glycine receptor alpha 2 subunit (glycine receptor, neonatal); Glycine receptor strychnine-binding subunit; Glycine receptor subunit alpha-2; glycine receptor, alpha 2; glycine receptor, alpha 2 subunit; glycine receptor, alpha-2 polypeptide
Gene Aliases: GLR; GLRA2; RNNEOGLY
UniProt ID: (Human) P23416, (Rat) P22771, (Mouse) Q7TNC8
Entrez Gene ID: (Human) 2742, (Rat) 24397, (Mouse) 237213
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.