Invitrogen
Manufacturer Code:PA521673
Catalog # PIPA521673
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 216 of Glycine Receptor alpha 2 (Uniprot IDNo.P23416) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GLR glycine receptor alpha 2 subunit glycine receptor subunit alpha-2 glycine receptor alpha-2 polypeptide; glycine receptor alpha 2 subunit; Glycine receptor subunit alpha-2; glycine receptor, alpha 2; glycine receptor, alpha-2 polypeptide
Gene Aliases: GLR; GLRA2
UniProt ID: (Human) P23416, (Mouse) Q7TNC8
Entrez Gene ID: (Human) 2742, (Mouse) 237213
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.