Abnova
Manufacturer Code:H00002741M03
Catalog # 89-022-324
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2B9 |
Immunogen | GLRA1 (NP_000162 121 a.a. ~ 220 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant GLRA1.
For Research Use Only
Protein Aliases: Glycine receptor 48 kDa subunit; Glycine receptor strychnine-binding subunit; Glycine receptor subunit alpha-1; MGC138878; MGC138879; STHE
Gene Aliases: GLRA1; HKPX1; STHE
UniProt ID: (Human) P23415
Entrez Gene ID: (Human) 2741
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.