Invitrogen
Manufacturer Code:PA577445
Catalog # PIPA577445
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)KRPDDESGWMSYLSE corresponding to amino acid residues 241-255 of rat GLP2R |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GLP-2 receptor; GLP-2-R; GLP-2R; GLP2R; Glucagon-like peptide 2 receptor; glucagon-like peptide-2 receptor
Gene Aliases: Glp2r
UniProt ID: (Rat) Q9Z0W0
Entrez Gene ID: (Rat) 60432
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.