Proteintech
Manufacturer Code:261961AP150UL
Catalog # 50-173-5252
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GLP1R Fusion Protein Ag23529 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GLP 1 R GLP 1 receptor GLP 1R GLP1R; GLP-1 receptor; GLP-1-R; GLP-1R; GLP1 receptor; Glucagon-like peptide 1 receptor; glucagon-like peptide-1 receptor; pancreatic beta cell receptor for the gluco-incretin hormone glucagon-like peptide 1; seven transmembrane helix receptor
Gene Aliases: Glip; GLP-1; GLP-1-R; GLP-1R; GLP1R; GLP1Rc; Glpr; RATGL1RCP
UniProt ID: (Human) P43220, (Mouse) O35659, (Rat) P32301
Entrez Gene ID: (Human) 2740, (Mouse) 14652, (Rat) 25051
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.