Novus Biologicals
Manufacturer Code:NBP223558JF646
Catalog # NBP23558646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4 |
Immunogen | Synthetic GLP-1(7-17) adsorbed onto aluminum hydroxide gel |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Glicentin; Glicentin-related polypeptide; glicentin-related polypeptide GLP1 glucagon-like peptide 1 GRPP; GLP-1; GLP-1(7-36); GLP-1(7-37); GLP-2; Glucagon; Glucagon-like peptide 1; Glucagon-like peptide 1(7-36); Glucagon-like peptide 1(7-37); Glucagon-like peptide 2; GRPP; Incretin hormone; OXM; Oxyntomodulin; preproglucagon; Pro-glucagon
Gene Aliases: GCG; GLP1; GLP2; GRPP
UniProt ID: (Human) P01275
Entrez Gene ID: (Human) 2641
Molecular Function: peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.