Novus Biologicals
Manufacturer Code:NBP177400F
Catalog # NBP177400F
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A genomic peptide made to an N-terminal region of the human EHMT1 protein (within residues 20-200). [Swiss-Prot Q9H9B1] |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.1.1.- EC 2.1.1.43 euchromatic histone-lysine N-methyltransferase 1H3-K9-HMTase 5 Eu-HMTase1DEL9q34 EUHMTASE1FP13812 FLJ12879 G9a-like protein 1 GLP GLP1 H3 lysine-9 specific 5 KIAA1876euchromatic histone methyltransferase 1 KMT1DDKFZp667M072; Eu-HMTase1; Euchromatic histone-lysine N-methyltransferase 1; G9a-like protein 1; GLP; H3-K9-HMTase 5; Histone H3-K9 methyltransferase 5; Histone-lysine N-methyltransferase EHMT1; histone-lysine N-methyltransferase, H3 lysine-9 specific 5; Lysine N-methyltransferase 1D
Gene Aliases: bA188C12.1; EHMT1; Eu-HMTase1; EUHMTASE1; FP13812; GLP; GLP1; KIAA1876; KMT1D
UniProt ID: (Human) Q9H9B1
Entrez Gene ID: (Human) 79813
Molecular Function: DNA binding protein methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.