R&D Systems
Manufacturer Code:FAB4285R100UG
Catalog # FAB4285R100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 413801 |
Immunogen | HEK293 human embryonic kidney cell line transfected with human GLP-2R Met1-Ile553 Accession # O95838 |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: GLP-2 receptor; GLP-2 receptor GLP2R GLP-2R GLP-2-R glucagon-like peptide 2 receptor; Glucagon-like peptide 2 receptor
Gene Aliases: GLP2R
UniProt ID: (Human) O95838
Entrez Gene ID: (Human) 9340
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.