Novus Biologicals
Manufacturer Code:NBP105062AF594
Catalog # NP10062A94
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1100000 |
Immunogen | Synthetic peptide coupled to diphtheria toxoid with glutaraldehyde |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: GLP-2 receptor; GLP-2 receptor GLP-2R GLP-2-R glucagon-like peptide 2 receptor; Glucagon-like peptide 2 receptor
Gene Aliases: GLP2R
UniProt ID: (Human) O95838
Entrez Gene ID: (Human) 9340
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.