R&D Systems
Manufacturer Code:FAB2814V100UG
Catalog # FAB2814V100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 197920 |
Immunogen | NS0 mouse myeloma cell line transfected with human GLP-1R Arg24-Ser463 Accession # NP_002053 |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: GLP-1 receptor; GLP-1 receptor GLP1R GLP-1R GLP-1-R glucagon-like peptide 1 receptor MGC138331; GLP1 receptor; Glucagon-like peptide 1 receptor; glucagon-like peptide-1 receptor; seven transmembrane helix receptor
Gene Aliases: GLP-1; GLP-1-R; GLP-1R; GLP1R
UniProt ID: (Human) P43220
Entrez Gene ID: (Human) 2740
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.