Novus Biologicals
Manufacturer Code:NBP197308R
Catalog # NBP197308R
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an internal portion of the human GLP1R protein (between residues 250-350) [UniProt P43220] |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GLP-1 receptor; GLP-1 receptor GLP-1R GLP-1-R glucagon-like peptide 1 receptor MGC138331; GLP1 receptor; Glucagon-like peptide 1 receptor; glucagon-like peptide-1 receptor; seven transmembrane helix receptor
Gene Aliases: GLP-1; GLP-1-R; GLP-1R; GLP1R
UniProt ID: (Human) P43220
Entrez Gene ID: (Human) 2740
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.