Invitrogen
Manufacturer Code:PA579315
Catalog # PIPA579315
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human GLO1 recombit protein (Position: A2-M184). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aldoketomutase; epididymis secretory protein Li 74; Glx I; glyoxalase 1 complex; glyoxalase 1 regulatory; glyoxalase 1 structural; glyoxalase domain containing 1; Glyoxalase I; glyoxylase 1; Ketone-aldehyde mutase; lactoyl glutathione lyase; Lactoylglutathione lyase; Lactoylglutathione lyase Aldoketomutase Glyoxalase I Glx I Ketone-aldehyde mutase Methylglyoxalase S-D-lactoylglutathione methylglyoxal lyase GLO1; Methylglyoxalase; S-D-lactoylglutathione methylglyoxal lyase
Gene Aliases: 0610009E22Rik; 1110008E19Rik; 2510049H23Rik; AW550643; Glo-1; Glo-1r; Glo-1s; GLO1; Glo1-r; Glo1-s; GLOD1; GLY1; GLYI; HEL-S-74; Qglo
UniProt ID: (Human) Q96J41, (Rat) Q6P7Q4, (Mouse) Q9CPU0
Entrez Gene ID: (Human) 2739, (Rat) 294320, (Mouse) 109801
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.