Invitrogen
Manufacturer Code:PA577987
Catalog # PIPA577987
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human Glyoxalase I. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aldoketomutase; epididymis secretory protein Li 74; GLOD1 GLYI GLO1 aldoketomutase glyoxalase domain containing 1 lactoylglutathione lyase S-D-lactoylglutathione methylglyoxal lyase lactoyl glutathione lyase methylglyoxalase ketone-aldehyde mutase glx I glyoxalase I; Glx I; glyoxalase 1 complex; glyoxalase 1 regulatory; glyoxalase 1 structural; glyoxalase domain containing 1; Glyoxalase I; Ketone-aldehyde mutase; lactoyl glutathione lyase; Lactoylglutathione lyase; Methylglyoxalase; S-D-lactoylglutathione methylglyoxal lyase
Gene Aliases: 0610009E22Rik; 1110008E19Rik; 2510049H23Rik; AW550643; Glo-1; Glo-1r; Glo-1s; GLO1; Glo1-r; Glo1-s; GLOD1; GLY1; GLYI; HEL-S-74; Qglo
UniProt ID: (Human) Q96J41, (Mouse) Q9CPU0
Entrez Gene ID: (Human) 2739, (Mouse) 109801
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.