Invitrogen
Manufacturer Code:PA588301
Catalog # PIPA588301
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 405-594 of human GLMN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FAP; FK506-binding protein-associated protein; FKBP-associated protein; Glomulin
Gene Aliases: 9330160J16Rik; AW227515; FAP; FAP48; FAP68; FKBPAP; GLML; GLMN; GVM; VMGLOM
UniProt ID: (Human) Q92990, (Mouse) Q8BZM1
Entrez Gene ID: (Human) 11146, (Mouse) 170823
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.