Invitrogen
Manufacturer Code:PA5103295
Catalog # PIPA5103295
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human GLI-3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACLS add AI854843 anterior digit pattern deformity AU023367 Bph brachyphalangy extra toes GCPS GLI family zinc finger 3 GLI family zinc finger 3 L homeolog gli3 GLI3 C-terminally truncated form GLI3 form of 190 kDa GLI3 form of 83 kDa GLI3 full length protein GLI3 full-length protein gli3.L GLI3-190 GLI3-83 GLI3FL GLI-Kruppel family member 3 GLI-Kruppel family member GLI3 GLI-Kruppel family member GLI3 (Greig cephalopolysyndactyly syndrome) glioma-associated oncogene family zinc finger 3 im:7142603 neural specific DNA binding protein neural-specific DNA-binding protein xGLI3 oncogene GLI3 PAPA PAP-A PAPA1 PAPB Pdn PHS polydactyly Nagoya PPDIV Transcriptional activator GLI3 Transcriptional repressor GLI3R XELAEV_18031191mg xgli3 xGLI-3 Xt Zinc finger protein GLI3 zinc finger transcription factor Gli3; anterior digit pattern deformity; brachyphalangy; extra toes; GLI-Kruppel family member GLI3; GLI3 C-terminally truncated form; GLI3 form of 190 kDa; GLI3 form of 83 kDa; GLI3 full length protein; GLI3 full-length protein; GLI3-190; GLI3-83; GLI3FL; glioma-associated oncogene family zinc finger 3; oncogene GLI3; polydactyly Nagoya; Transcriptional activator GLI3; Transcriptional repressor GLI3R; zinc finger protein GLI3
Gene Aliases: ACLS; add; AI854843; AU023367; Bph; GCPS; GLI3; GLI3-190; GLI3FL; PAP-A; PAPA; PAPA1; PAPB; Pdn; PHS; PPDIV; Xt
UniProt ID: (Human) P10071, (Mouse) Q61602
Entrez Gene ID: (Human) 2737, (Rat) 140588, (Mouse) 14634
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.