Abnova
Manufacturer Code:H00002737M01
Catalog # 89-019-025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2C9 |
Immunogen | GLI3 (NP_000159 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant GLI3.
For Research Use Only
Protein Aliases: ACLS; GCPS; GLI-Kruppel family member GLI3; GLI3 C-terminally truncated form; GLI3 form of 190 kDa; GLI3 form of 83 kDa; GLI3 full-length protein; GLI3-190; GLI3-83; GLI3FL; glioma-associated oncogene family zinc finger 3; oncogene GLI3; PAP-A; PAPA; PAPA1; PAPB; PHS; PPDIV; Transcriptional activator GLI3; Transcriptional repressor GLI3R; zinc finger protein GLI3
Gene Aliases: ACLS; GCPS; GLI3; GLI3-190; GLI3FL; PAP-A; PAPA; PAPA1; PAPB; PHS; PPDIV
UniProt ID: (Human) P10071
Entrez Gene ID: (Human) 2737
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.