Novus Biologicals
Manufacturer Code:NBP223602AF594
Catalog # NP223602A94
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an internal portion of the human GLI-2 protein (between residues 300-400) [UniProtNo. P10070] |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
Protein Aliases: GLI family zinc finger 2 GLI-Kruppel family member GLI2 glioma-associated oncogene family zinc finger 2 HPE9tax helper protein 1 oncogene GLI2 Tax helper protein tax helper protein 2 tax-responsive element-2 holding protein tax-responsive element-25-bp sequence binding protein THP THP1 THP2 zinc finger protein GLI2; GLI family zinc finger protein 2; GLI-Kruppel family member GLI2; glioma-associated oncogene family zinc finger 2; oncogene GLI2; Tax helper protein; tax helper protein 1; tax helper protein 2; tax-responsive element-2 holding protein; tax-responsive element-25-bp sequence binding protein; Zinc finger protein GLI2
Gene Aliases: CJS; GLI2; HPE9; PHS2; THP; THP1; THP2
UniProt ID: (Human) P10070
Entrez Gene ID: (Human) 2736
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.