R&D Systems
Manufacturer Code:AF3526SP
Catalog # AF3526SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human GLI-2 Ala2-Glu91 Accession # BAA25668 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: GLI family zinc finger 2 GLI2 GLI-Kruppel family member GLI2 glioma-associated oncogene family zinc finger 2 HPE9tax helper protein 1 oncogene GLI2 Tax helper protein tax helper protein 2 tax-responsive element-2 holding protein tax-responsive element-25-bp sequence binding protein THP THP1 THP2 zinc finger protein GLI2; GLI family zinc finger protein 2; GLI-Kruppel family member GLI2; glioma-associated oncogene family zinc finger 2; oncogene GLI2; Tax helper protein; tax helper protein 1; tax helper protein 2; tax-responsive element-2 holding protein; tax-responsive element-25-bp sequence binding protein; Zinc finger protein GLI2
Gene Aliases: CJS; GLI2; HPE9; PHS2; THP; THP1; THP2
UniProt ID: (Human) P10070
Entrez Gene ID: (Human) 2736
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.