Abnova
Manufacturer Code:PAB12523
Catalog # 89-115-898
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 160-176 of human GLDN. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for short term. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of GLDN.
For Research Use Only
Protein Aliases: CLOM; collomin; COLM; colmedin; CRG-L2; CRGL2; FLJ23917; Gliomedin; Gliomedin shedded ectodomain; UNC-112
Gene Aliases: CLOM; COLM; CRG-L2; CRGL2; GLDN; UNC-112; UNQ9339/PRO34011
UniProt ID: (Human) Q6ZMI3
Entrez Gene ID: (Human) 342035
Molecular Function: receptor structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.