Proteintech
Manufacturer Code:261851AP150UL
Catalog # 50-173-5246
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GLDN Fusion Protein Ag24247 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cancer related gene - liver 2; Cancer-related gene liver 2 protein; CLOM COLM CRG L2 CRGL2 GLDN gliomedin UNC 112; collomin; colmedin; CRG-L2; Gliomedin; Gliomedin shedded ectodomain; olfactomedin-related protein
Gene Aliases: CLOM; COLM; CRG-L2; CRGL2; Crlg2; GLDN; UNC-112; UNQ9339/PRO34011
UniProt ID: (Human) Q6ZMI3, (Mouse) Q8BMF8
Entrez Gene ID: (Human) 342035, (Mouse) 235379
Molecular Function: receptor structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.