Invitrogen
Manufacturer Code:PA597294
Catalog # PIPA597294
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 36-290 of human GLDC. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glycine cleavage system P protein; glycine cleavage system protein P; Glycine decarboxylase; glycine decarboxylase P-protein; Glycine dehydrogenase (aminomethyl-transferring); glycine dehydrogenase (decarboxylating); Glycine dehydrogenase (decarboxylating), mitochondrial; glycine dehydrogenase [decarboxylating], mitochondrial
Gene Aliases: D030049L12Rik; D19Wsu57e; GCE; GCSP; GLDC; HYGN1
UniProt ID: (Human) P23378, (Mouse) Q91W43
Entrez Gene ID: (Human) 2731, (Mouse) 104174
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.