Invitrogen
Manufacturer Code:PA551701
Catalog # PIPA551701
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human GLDC |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glycine cleavage system P protein; glycine cleavage system protein P; Glycine decarboxylase; glycine decarboxylase P-protein; glycine decarboxylase, glycine cleavage system protein P); Glycine dehydrogenase (aminomethyl-transferring); glycine dehydrogenase (decarboxylating); Glycine dehydrogenase (decarboxylating), mitochondrial; glycine dehydrogenase (decarboxylating; glycine decarboxylase, glycine cleavage system protein P); glycine dehydrogenase [decarboxylating], mitochondrial; RP11-472F14.1 GCE GCSP HYGN1 glycine cleavage system P protein glycine cleavage system protein P glycine decarboxylase P-protein glycine dehydrogenase &lsqbdecarboxylating] mitochondrial
Gene Aliases: D030049L12Rik; D19Wsu57e; GCE; GCSP; GLDC; HYGN1
UniProt ID: (Human) P23378, (Mouse) Q91W43
Entrez Gene ID: (Human) 2731, (Mouse) 104174, (Rat) 309312
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.