Novus Biologicals
Manufacturer Code:NBP304989100UL
Catalog # NB092745
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 36-290 of human GLDC (NP_000161.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 1.4.4.2 GCE GCSPMGC138198 Glycine cleavage system P protein glycine cleavage system protein P Glycine decarboxylase glycine decarboxylase P-protein glycine dehydrogenase (decarboxylating) glycine dehydrogenase (decarboxylating glycine decarboxylase glycine cleavagesystem protein P) glycine dehydrogenase [decarboxylating] mitochondrial HYGN1 MGC138200 NKH; Glycine cleavage system P protein; glycine cleavage system protein P; Glycine decarboxylase; glycine decarboxylase P-protein; Glycine dehydrogenase (aminomethyl-transferring); glycine dehydrogenase (decarboxylating); Glycine dehydrogenase (decarboxylating), mitochondrial; glycine dehydrogenase [decarboxylating], mitochondrial
Gene Aliases: GCE; GCSP; GLDC; HYGN1
UniProt ID: (Human) P23378
Entrez Gene ID: (Human) 2731
Molecular Function:
dehydrogenase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.