Abnova
Manufacturer Code:PAB21084
Catalog # 89-122-580
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human GLDC. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant GLDC.
For Research Use Only
Protein Aliases: GCE; GCSP; Glycine cleavage system P protein; glycine cleavage system protein P; Glycine decarboxylase; glycine decarboxylase P-protein; Glycine dehydrogenase (aminomethyl-transferring); glycine dehydrogenase (decarboxylating); Glycine dehydrogenase (decarboxylating), mitochondrial; glycine dehydrogenase [decarboxylating], mitochondrial; HYGN1; MGC138198; MGC138200; NKH
Gene Aliases: GCE; GCSP; GLDC; HYGN1
UniProt ID: (Human) P23378
Entrez Gene ID: (Human) 2731
Molecular Function: dehydrogenase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.