Invitrogen
Manufacturer Code:PA561688
Catalog # PIPA561688
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human GLCE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D-glucuronyl C5-epimerase; glucuronyl C5-epimerase; Heparan sulfate C5-epimerase; heparan sulfate epimerase; heparin/heparan sulfate-glucuronic acid C5-epimerase; Heparin/heparan sulfate:glucuronic acid C5-epimerase; Heparosan-N-sulfate-glucuronate 5-epimerase; Hsepi; HSEPI D-glucuronyl C5-epimerase UDP-glucuronic acid epimerase glucuronyl C5-epimerase heparan sulfate epimerase heparin/heparan sulfate-glucuronic acid C5-epimerase heparin/heparan sulfate:glucuronic acid C5-epimerase; UDP-glucuronic acid epimerase
Gene Aliases: GLCE; HSEPI; KIAA0836
UniProt ID: (Human) O94923
Entrez Gene ID: (Human) 26035
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.