Invitrogen
Manufacturer Code:PA575874
Catalog # PIPA575874
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human GLB1L3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-galactosidase-1-like protein 3; Beta-galactosidase-1-like protein 3 (EC:3.2.1.-) GLB1L3; galactosidase beta 1-like 3; galactosidase, beta 1 like 3; galactosidase, beta 1-like 3
Gene Aliases: 4921509F24Rik; GLB1L3
UniProt ID: (Human) Q8NCI6, (Mouse) A2RSQ1, (Rat) Q5XIL5
Entrez Gene ID: (Human) 112937, (Mouse) 70893, (Rat) 500961
Molecular Function: galactosidase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.