Invitrogen
Manufacturer Code:PA572071
Catalog # PIPA572071
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 123-150 amino acids from the N-terminal region of human GLB1L |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-galactosidase-1-like protein; Beta-galactosidase-1-like protein 321- GLB1L; galactosidase beta 1-like; galactosidase, beta 1-like; testicular tissue protein Li 74
Gene Aliases: 4833408P15Rik; GLB1L; UNQ229/PRO262
UniProt ID: (Human) Q6UWU2, (Mouse) Q8VC60
Entrez Gene ID: (Human) 79411, (Mouse) 74577
Molecular Function: galactosidase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.