Invitrogen
Manufacturer Code:MA526152
Catalog # PIMA526152
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1C9 |
Immunogen | Full length human recombit protein of GLB1 produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Acid beta galactosidaseAcid beta-galactosidaseBeta galactosidase 1Beta galactosidaseBeta-galactosidaseBGAL&lowbarHUMANEBPEBP includedElastin receptor 1 (67kD)Elastin receptor 1 67kDaElastin receptor 1Elastin receptor 1 includedElastin-binding protein includedELNR1Galactosidase beta 1GLB 1GLB1LactaseMPS4BS-GAL included; Acid beta-galactosidase; Beta-galactosidase; Elastin receptor 1; elastin receptor 1, 67kDa; GLB1; Lactase; lysosmal hydrolase; lysosomal beta-galactosidase; lysosomal hydrolase; unnamed protein product
Gene Aliases: EBP; ELNR1; GLB1; Lactase; MPS4B; QccE-15801; QccE-17236
UniProt ID: (Human) P16278, (Cynomolgus monkey) Q60HF6, (Dog) Q9TRY9
Entrez Gene ID: (Human) 2720, (Cynomolgus monkey) 102119163, (Dog) 403873
Molecular Function:
galactosidase
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.