Invitrogen
Manufacturer Code:PA519709
Catalog # PIPA519709
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide conjugated to KLH derived from within residues 1 - 100 of Human EAAT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EA6 EAAT1 FLJ25094 GLAST GLAST1 GLAST-1 excitatory amino acid transporter 1 sodium-dependent glutamate/aspartate transporter 1 solute carrier family 1 member 3; Excitatory amino acid transporter 1; GLAST; GLAST-1; Glial glutamate transporter; Glial high affinity glutamate transporter; GluT-1; glutamate transporter; glutamate/aspartate transporter; High-affinity neuronal glutamate transporter; Sodium-dependent glutamate/aspartate transporter 1; solute carrier family 1 (glial high affinity glutamate transporter), member 3; Solute carrier family 1 member 3; solute carrier family 1, member 3
Gene Aliases: AI504299; B430115D02Rik; EA6; EAAT1; GLAST; GLAST-1; GLAST1; GLU-T; GluT-1; Gmt1; MGluT1; SLC1A3
UniProt ID: (Human) P43003, (Rat) P24942, (Mouse) P56564
Entrez Gene ID: (Human) 6507, (Rat) 29483, (Mouse) 20512
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.