Invitrogen
Manufacturer Code:PA560045
Catalog # PIPA560045
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human GK5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP glycerol 3-phosphotransferase 5 GK 5 glycerokinase 5 putative glycerol kinase 5; ATP:glycerol 3-phosphotransferase 5; GK 5; glycerokinase 5; Putative glycerol kinase 5
Gene Aliases: GK5
UniProt ID: (Human) Q6ZS86
Entrez Gene ID: (Human) 256356
Molecular Function: carbohydrate kinase kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.