Proteintech
Manufacturer Code:134571AP150UL
Catalog # 50-172-8759
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GK5 Fusion Protein Ag4269 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP:glycerol 3-phosphotransferase 5; GK 5; GK 5 GK5 Glycerokinase 5 glycerol kinase 5 (putative) Putative glycerol kinase 5; glycerokinase 5; glycerol kinase 5 (putative); Putative glycerol kinase 5
Gene Aliases: AV095337; C330018K18Rik; G630067D24Rik; GK5; RGD1311223
UniProt ID: (Human) Q6ZS86, (Mouse) Q8BX05
Entrez Gene ID: (Human) 256356, (Mouse) 235533, (Rat) 367146
Molecular Function:
carbohydrate kinase
kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.