Novus Biologicals
Manufacturer Code:NBP281089AF350
Catalog # NB015795
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | YGL386 |
Immunogen | Recombinant protein injected into DA rats. |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Activation-inducible TNF-related ligand; Activation-inducible TNF-related ligand AITRLMGC138237 GITRLglucocorticoid-induced TNFR-related protein ligand Glucocorticoid-induced TNF-related ligand hGITRLGITR ligand TL6AITR ligand tumor necrosis factor (ligand) superfamily member 18 tumor necrosis factor ligand superfamily member 18; AITR ligand; AITRL; GITR ligand; Glucocorticoid-induced TNF-related ligand; glucocorticoid-induced TNFR-related protein ligand; hGITRL; tumor necrosis factor (ligand) superfamily member 18; tumor necrosis factor (ligand) superfamily, member 18; tumor necrosis factor ligand 2A; Tumor necrosis factor ligand superfamily member 18
Gene Aliases: AITRL; GITRL; hGITRL; TL6; TNFSF18; TNLG2A; UNQ149/PRO175
UniProt ID: (Human) Q9UNG2
Entrez Gene ID: (Human) 8995
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.