R&D Systems
Manufacturer Code:MAB21772SP
Catalog # MAB21772SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 721926 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse GITR Ligand/TNFSF18 Thr47-Ser173 Accession # NP_899247 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Activation-inducible TNF-related ligand; Activation-inducible TNF-related ligand AITRL AITRLMGC138237 GITRL GITRLglucocorticoid-induced TNFR-related protein ligand Glucocorticoid-induced TNF-related ligand hGITRLGITR ligand TL6AITR ligand TNFSF18 tumor necrosis factor (ligand) superfamily member 18 tumor necrosis factor ligand superfamily member 18; AITR ligand; AITRL; GITR ligand; Glucocorticoid-induced TNF-related ligand; glucocorticoid-induced TNFR-related protein ligand; hGITRL; tumor necrosis factor (ligand) superfamily member 18; tumor necrosis factor (ligand) superfamily, member 18; tumor necrosis factor ligand 2A; Tumor necrosis factor ligand superfamily member 18
Gene Aliases: AITRL; GITRL; hGITRL; TL6; TNFSF18; TNLG2A; UNQ149/PRO175
UniProt ID: (Human) Q9UNG2
Entrez Gene ID: (Human) 8995
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.