Proteintech
Manufacturer Code:216471AP150UL
Catalog # 50-173-3534
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GIRK2 Fusion Protein Ag16344 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BIR1; BIR1 GIRK 2 GIRK2 hiGIRK2 KATP 2 KATP2 KCNJ6 KCNJ7 KIR3.2; G protein-activated inward rectifier potassium channel 2; GIRK-2; GIRK2; Inward rectifier K(+) channel Kir3.2; inward rectifier potassium channel KIR3.2; KATP-2; Potassium channel, inwardly rectifying subfamily J member 6; potassium channel, inwardly rectifying subfamily J, member 6; potassium inwardly-rectifying channel J6; potassium inwardly-rectifying channel, subfamily J, member 6
Gene Aliases: BIR1; GIRK-2; GIRK2; hiGIRK2; KATP-2; KATP2; KCNJ6; KCNJ7; KIR3.2; KPLBS; W; weaver; wv
UniProt ID: (Human) P48051, (Mouse) P48542, (Rat) P48550
Entrez Gene ID: (Human) 3763, (Mouse) 16522, (Rat) 25743
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.