Invitrogen
Manufacturer Code:PA577447
Catalog # PIPA577447
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)EGQTTGELYQRWER corresponding to amino acid residues 25-38 of rat Gastric Inhibitory Polypeptide Receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: gastric inhibitory peptide receptor; Gastric inhibitory polypeptide receptor; Gastric Inhibitory Polypeptide Receptor (extracellular); GIP-R; Glucose-dependent insulinotropic polypeptide receptor
Gene Aliases: GIP-R; Gippr; Gipr; Gm1081; Gm160; RATGIPPR
UniProt ID: (Mouse) Q0P543, (Rat) P43219
Entrez Gene ID: (Mouse) 381853, (Rat) 25024
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.