Novus Biologicals
Manufacturer Code:MAB82101AF532
Catalog # MB82101A532
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 591816 |
Immunogen | NS0 mouse myeloma cell line transfected with human GIPR (amino acids 1-466) |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Gastric inhibitory polypeptide receptor; gastric inhibitory polypeptide receptor GIP-R Glucose-dependent insulinotropic polypeptide receptor MGC126722 PGQTL2; GIP-R; Glucose-dependent insulinotropic polypeptide receptor
Gene Aliases: GIPR; PGQTL2
UniProt ID: (Human) P48546
Entrez Gene ID: (Human) 2696
Molecular Function:
G-protein coupled receptor
antibacterial response protein
defense/immunity protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.