Invitrogen
Manufacturer Code:PA576437
Catalog # PIPA576437
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human GIPR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: gastric inhibitory peptide receptor; Gastric inhibitory polypeptide receptor; Gastric inhibitory polypeptide receptor GIP-R; GIP-R; Glucose-dependent insulinotropic polypeptide receptor
Gene Aliases: GIP-R; Gippr; GIPR; Gm1081; Gm160; PGQTL2; RATGIPPR
UniProt ID: (Human) P48546, (Mouse) Q0P543, (Rat) P43219
Entrez Gene ID: (Human) 2696, (Mouse) 381853, (Rat) 25024
Molecular Function:
G-protein coupled receptor
antibacterial response protein
defense/immunity protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.