Novus Biologicals
Manufacturer Code:NBP221617
Catalog # NBP221617
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4 |
Immunogen | Synthetic human gastric inhibitory polypeptide adsorbed onto aluminum hydroxide gel. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Gastric inhibitory polypeptide; gastric inhibitory polypeptide Glucose-dependent insulinotropic polypeptide; GIP; Glucose-dependent insulinotropic polypeptide; Incretin hormone
Gene Aliases: GIP
UniProt ID: (Human) P09681
Entrez Gene ID: (Human) 2695
Molecular Function: peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.