Invitrogen
Manufacturer Code:PA571571
Catalog # PIPA571571
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 464-492 amino acids from the C-terminal region of human GIN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GIN-1; Gypsy retrotransposon integrase-like protein 1; Gypsy retrotransposon integrase-like protein 1 GIN-1 Ty3/Gypsy integrase 1 Zinc finger H2C2 domain-containing protein GIN1 TGIN1 ZH2C2; Zinc finger H2C2 domain-containing protein; zinc finger, H2C2 domain containing
Gene Aliases: 4930429M06Rik; 8430411H10Rik; Gin-1; Gin1; Zh2c2
UniProt ID: (Mouse) Q8K259
Entrez Gene ID: (Mouse) 252876
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.