Invitrogen
Manufacturer Code:PA566381
Catalog # PIPA566381
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human GIN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GIN-1; Gypsy retrotransposon integrase-like protein 1; TGIN1 ZH2C2; Ty3/Gypsy integrase 1; Zinc finger H2C2 domain-containing protein
Gene Aliases: GIN-1; GIN1; TGIN1; ZH2C2
UniProt ID: (Human) Q9NXP7
Entrez Gene ID: (Human) 54826
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.