Invitrogen
Manufacturer Code:PA545586
Catalog # PIPA545586
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human GIGYF2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GRB10 interacting GYF protein 2; GRB10-interacting GYF protein 2; Parkinson disease (autosomal recessive, early onset) 11; PERQ amino acid rich, with GYF domain 3; PERQ amino acid-rich with GYF domain-containing 2-like protein; PERQ amino acid-rich with GYF domain-containing protein 2; rCG23949-like; trinucleotide repeat containing 15; Trinucleotide repeat-containing gene 15 protein
Gene Aliases: 2610016F01Rik; A830080H02Rik; AI852361; AW259676; BC006835; GIGYF2; GYF2; KIAA0642; mKIAA0642; PARK11; PERQ2; PERQ3; TNRC15
UniProt ID: (Human) Q6Y7W6, (Mouse) Q6Y7W8
Entrez Gene ID: (Human) 26058, (Bovine) 506913, (Dog) 612838, (Horse) 100064814, (Rabbit) 100348353, (Guinea pig) 100732403, (Mouse) 227331, (Rat) 100362458
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.