Novus Biologicals
Manufacturer Code:NBP29022550UL
Catalog # NB021914
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 7 |
Immunogen | Recombinant Human GIF Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: Cobalamin binding intrinsic factor; Gastric intrinsic factor; gastric intrinsic factor (vitamin B synthesis); gastric intrinsic factor (vitamin B synthesis) IFIntrinsic factor IFMHgastric intrinsic factor INF TCN3; IF; Intrinsic factor
Gene Aliases: CBLIF; GIF; IF; IFMH; INF; TCN3
UniProt ID: (Human) P27352
Entrez Gene ID: (Human) 2694
Molecular Function: cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.